Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35074077-35074328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
UniGene
Hs.591084
Gene Chromosome Location
Chr.9: 35073838-35080016 on Build GRCh38
Transcripts
NM_004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4401446 c.1807T>A p.S603T Chr.9: 35074170-35074170 on Build GRCh38
3699672 c.1768C>A p.P590T Chr.9: 35074209-35074209 on Build GRCh38
5559782 c.1766T>C p.L589P Chr.9: 35074211-35074211 on Build GRCh38
1462189 c.1821C>A p.A607A Chr.9: 35074156-35074156 on Build GRCh38
6036225 c.1761-8C>A p.? Chr.9: 35074224-35074224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541868979 Chr.9: 35074209-35074209 on Build GRCh38
rs374723194 Chr.9: 35074175-35074175 on Build GRCh38
rs771669019 Chr.9: 35074112-35074112 on Build GRCh38
rs773844815 Chr.9: 35074104-35074104 on Build GRCh38
rs772995473 Chr.9: 35074219-35074219 on Build GRCh38
rs370950631 Chr.9: 35074172-35074172 on Build GRCh38
rs766323286 Chr.9: 35074233-35074233 on Build GRCh38
rs748634605 Chr.9: 35074139-35074139 on Build GRCh38
rs768325047 Chr.9: 35074203-35074203 on Build GRCh38
rs17878854 Chr.9: 35074169-35074169 on Build GRCh38
rs763852498 Chr.9: 35074127-35074127 on Build GRCh38
rs756632525 Chr.9: 35074171-35074171 on Build GRCh38
rs928814452 Chr.9: 35074166-35074166 on Build GRCh38
rs760359166 Chr.9: 35074123-35074123 on Build GRCh38
rs746392434 Chr.9: 35074206-35074206 on Build GRCh38
rs562165828 Chr.9: 35074237-35074237 on Build GRCh38
rs949414610 Chr.9: 35074192-35074192 on Build GRCh38
rs572470999 Chr.9: 35074254-35074254 on Build GRCh38
rs770455405 Chr.9: 35074101-35074101 on Build GRCh38
rs928548559 Chr.9: 35074245-35074245 on Build GRCh38
rs150203467 Chr.9: 35074136-35074136 on Build GRCh38
rs1036849087 Chr.9: 35074278-35074278 on Build GRCh38
rs761695914 Chr.9: 35074140-35074140 on Build GRCh38
rs766524415 Chr.9: 35074159-35074159 on Build GRCh38
rs756542501 Chr.9: 35074260-35074260 on Build GRCh38
rs748619052 Chr.9: 35074100-35074100 on Build GRCh38
rs765150956 Chr.9: 35074218-35074218 on Build GRCh38
rs750436883 Chr.9: 35074157-35074157 on Build GRCh38
rs1026560078 Chr.9: 35074303-35074303 on Build GRCh38
rs776337442 Chr.9: 35074204-35074204 on Build GRCh38
rs370246800 Chr.9: 35074244-35074244 on Build GRCh38
rs397507560 Chr.9: 35074174-35074183 on Build GRCh38
rs771451437 Chr.9: 35074183-35074183 on Build GRCh38
rs758407400 Chr.9: 35074158-35074158 on Build GRCh38
rs541582645 Chr.9: 35074302-35074302 on Build GRCh38
rs775113723 Chr.9: 35074195-35074195 on Build GRCh38
rs755275747 Chr.9: 35074164-35074164 on Build GRCh38
rs532302967 Chr.9: 35074124-35074125 on Build GRCh38
rs758980894 Chr.9: 35074111-35074111 on Build GRCh38
rs763000489 Chr.9: 35074220-35074220 on Build GRCh38
rs373796469 Chr.9: 35074264-35074264 on Build GRCh38
rs568359829 Chr.9: 35074273-35074273 on Build GRCh38
rs749733793 Chr.9: 35074267-35074267 on Build GRCh38
rs751710424 Chr.9: 35074163-35074163 on Build GRCh38
rs898364396 Chr.9: 35074283-35074283 on Build GRCh38
rs765086974 Chr.9: 35074155-35074155 on Build GRCh38
rs746613103 Chr.9: 35074200-35074200 on Build GRCh38
rs775000011 Chr.9: 35074117-35074117 on Build GRCh38
rs368098479 Chr.9: 35074176-35074176 on Build GRCh38
rs901236235 Chr.9: 35074279-35074279 on Build GRCh38

View Full Product Details