The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130417.2 | NP_001123889.1 | 4 - 5 | 577 | 72 |
NM_033262.4 | NP_150287.1 | 6 - 7 | 2874 | 72 | |
NM_058240.3 | NP_489479.1 | 6 - 7 | 2871 | 72 | |
NM_182932.2 | NP_891977.1 | 5 - 6 | 2862 | 72 | |
NM_182936.2 | NP_891981.1 | 3 - 4 | 473 | 72 | |
NM_183002.2 | NP_892114.1 | 6 - 7 | 2880 | 72 | |
NR_104122.1 | 4 - 5 | 2758 | 72 | ||
XM_006720240.3 | XP_006720303.1 | 5 - 6 | 1726 | 72 | |
XM_017021606.1 | XP_016877095.1 | 6 - 7 | 2252 | 72 | |
XM_017021608.1 | XP_016877097.1 | 5 - 6 | 2488 | 72 | |
XM_017021611.1 | XP_016877100.1 | 4 - 5 | 850 | 72 | |
GenBank mRNA | AF510501.1 | - | 5 - 6 | 2862 | 72 |
AF510502.1 | - | 6 - 7 | 2880 | 72 | |
AF510503.1 | - | 4 - 5 | 2758 | 72 | |
AJ304852.1 | - | 5 - 6 | 2182 | 72 | |
AJ304853.1 | - | 5 - 6 | 2179 | 72 | |
AJ508602.1 | - | 4 - 5 | 1658 | 72 | |
AJ745101.1 | - | 3 - 4 | 436 | 72 | |
AJ745102.1 | - | 4 - 5 | 540 | 72 | |
AK096553.1 | - | 3 - 4 | 403 | 72 | |
AK122728.1 | - | 4 - 5 | 564 | 72 | |
BC036783.1 | - | 5 - 6 | 2665 | 72 | |
BC142969.1 | - | 4 - 5 | 566 | 72 | |
BC150183.1 | - | 4 - 5 | 2185 | 72 | |
BC150193.1 | - | 4 - 5 | 2185 | 72 |