Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42752896-42753141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3886995 c.2197C>T p.Q733* Chr.15: 42753049-42753049 on Build GRCh38
73105 c.2254C>G p.Q752E Chr.15: 42752992-42752992 on Build GRCh38
4535264 c.2173G>A p.G725R Chr.15: 42753073-42753073 on Build GRCh38
4846942 c.2335G>C p.E779Q Chr.15: 42752911-42752911 on Build GRCh38
6244700 c.2150C>A p.T717K Chr.15: 42753096-42753096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183390147 Chr.15: 42753030-42753030 on Build GRCh38
rs367778872 Chr.15: 42753089-42753089 on Build GRCh38
rs767399781 Chr.15: 42752955-42752955 on Build GRCh38
rs374412910 Chr.15: 42753080-42753080 on Build GRCh38
rs371333858 Chr.15: 42753032-42753032 on Build GRCh38
rs76000738 Chr.15: 42753113-42753113 on Build GRCh38
rs751381521 Chr.15: 42752951-42752951 on Build GRCh38
rs752400040 Chr.15: 42752947-42752947 on Build GRCh38
rs187389348 Chr.15: 42753104-42753104 on Build GRCh38
rs558836276 Chr.15: 42752977-42752977 on Build GRCh38
rs759376111 Chr.15: 42753071-42753071 on Build GRCh38
rs778756599 Chr.15: 42753115-42753115 on Build GRCh38
rs546749426 Chr.15: 42752936-42752936 on Build GRCh38
rs554361140 Chr.15: 42753056-42753056 on Build GRCh38
rs761835191 Chr.15: 42753051-42753051 on Build GRCh38
rs544778041 Chr.15: 42752921-42752921 on Build GRCh38
rs752644926 Chr.15: 42753085-42753085 on Build GRCh38
rs756796072 Chr.15: 42753114-42753114 on Build GRCh38
rs146515654 Chr.15: 42753036-42753036 on Build GRCh38
rs374595367 Chr.15: 42752986-42752986 on Build GRCh38
rs117382379 Chr.15: 42752968-42752968 on Build GRCh38
rs371090315 Chr.15: 42752965-42752965 on Build GRCh38
rs756990473 Chr.15: 42752983-42752983 on Build GRCh38
rs775916088 Chr.15: 42753009-42753009 on Build GRCh38
rs746274645 Chr.15: 42752997-42752997 on Build GRCh38
rs776707830 Chr.15: 42753037-42753037 on Build GRCh38
rs190887768 Chr.15: 42752953-42752953 on Build GRCh38
rs776781931 Chr.15: 42752930-42752930 on Build GRCh38
rs773395659 Chr.15: 42753057-42753057 on Build GRCh38
rs201524659 Chr.15: 42753075-42753075 on Build GRCh38

View Full Product Details