Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46645789-46646062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1980722 c.229G>A p.G77R Chr.11: 46645948-46645948 on Build GRCh38
6606600 c.238A>G p.M80V Chr.11: 46645957-46645957 on Build GRCh38
5360475 c.165C>A p.I55I Chr.11: 46645884-46645884 on Build GRCh38
1980723 c.229G>A p.G77R Chr.11: 46645948-46645948 on Build GRCh38
6606601 c.238A>G p.M80V Chr.11: 46645957-46645957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763447202 Chr.11: 46645825-46645825 on Build GRCh38
rs937938001 Chr.11: 46645881-46645881 on Build GRCh38
rs907758778 Chr.11: 46645980-46645980 on Build GRCh38
rs150268260 Chr.11: 46645965-46645965 on Build GRCh38
rs371039787 Chr.11: 46645943-46645943 on Build GRCh38
rs746329828 Chr.11: 46645826-46645826 on Build GRCh38
rs759249448 Chr.11: 46645905-46645905 on Build GRCh38
rs769242909 Chr.11: 46645861-46645861 on Build GRCh38
rs772737570 Chr.11: 46645866-46645866 on Build GRCh38
rs201305089 Chr.11: 46646022-46646022 on Build GRCh38
rs147713250 Chr.11: 46645938-46645938 on Build GRCh38
rs141220159 Chr.11: 46645849-46645849 on Build GRCh38
rs772654146 Chr.11: 46646036-46646036 on Build GRCh38
rs141061538 Chr.11: 46645947-46645947 on Build GRCh38
rs989751823 Chr.11: 46645882-46645882 on Build GRCh38
rs200971376 Chr.11: 46645822-46645822 on Build GRCh38
rs747842964 Chr.11: 46645856-46645856 on Build GRCh38
rs945171519 Chr.11: 46645925-46645925 on Build GRCh38
rs747756953 Chr.11: 46646007-46646007 on Build GRCh38
rs202100011 Chr.11: 46645852-46645852 on Build GRCh38
rs376130493 Chr.11: 46645848-46645848 on Build GRCh38
rs766243483 Chr.11: 46645886-46645886 on Build GRCh38
rs757578336 Chr.11: 46645936-46645936 on Build GRCh38
rs767387043 Chr.11: 46645911-46645911 on Build GRCh38
rs926558533 Chr.11: 46645831-46645831 on Build GRCh38
rs774409200 Chr.11: 46645894-46645894 on Build GRCh38
rs899009388 Chr.11: 46646021-46646021 on Build GRCh38
rs762884295 Chr.11: 46645878-46645878 on Build GRCh38
rs556193895 Chr.11: 46645928-46645928 on Build GRCh38
rs780406761 Chr.11: 46645842-46645842 on Build GRCh38
rs755271563 Chr.11: 46646001-46646001 on Build GRCh38
rs758832066 Chr.11: 46645833-46645833 on Build GRCh38
rs200546929 Chr.11: 46645906-46645906 on Build GRCh38
rs781555505 Chr.11: 46646006-46646006 on Build GRCh38
rs747591428 Chr.11: 46645998-46645998 on Build GRCh38
rs927088889 Chr.11: 46645887-46645887 on Build GRCh38
rs934021972 Chr.11: 46645996-46645996 on Build GRCh38
rs750542688 Chr.11: 46645944-46645944 on Build GRCh38
rs754091375 Chr.11: 46645929-46645929 on Build GRCh38

View Full Product Details