The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009186.1 | NP_001009186.1 | 5 - 6 | 759 | 102 |
NM_001762.3 | NP_001753.1 | 6 - 7 | 894 | 102 | |
GenBank mRNA | AB063318.1 | - | 6 - 7 | 866 | 102 |
AB209728.1 | - | 6 - 7 | 726 | 102 | |
AF385084.1 | - | 6 - 7 | 785 | 102 | |
AK297751.1 | - | 5 - 6 | 579 | 102 | |
AK298371.1 | - | 7 - 8 | 659 | 102 | |
AK313819.1 | - | 6 - 7 | 752 | 102 | |
AK316230.1 | - | 7 - 8 | 712 | 102 | |
AM295155.1 | - | 6 - 7 | 705 | 102 | |
BC106941.1 | - | 6 - 7 | 810 | 102 | |
BC106942.1 | - | 6 - 7 | 810 | 102 | |
BU540578.1 | - | 5 - 6 | 669 | 102 | |
L27706.1 | - | 6 - 7 | 816 | 102 | |
M94083.1 | - | 4 - 5 | 465 | 102 |