Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPA1P6
Entrez Gene ID
729423
Gene Name
heterogeneous nuclear ribonucleoprotein A1 pseudogene 6
Gene Aliases
HNRNPA1L, HNRNPA1L1, HNRPA1L
Chromosome Location
Chr.1: 58047849 - 58049373 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC121134.1 - 1 - 1 160 89

View Full Product Details