The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017818.3 | NP_060288.3 | 5 - 6 | 625 | 62 |
XM_005244754.1 | XP_005244811.1 | 5 - 6 | 620 | 62 | |
XM_017001387.1 | XP_016856876.1 | 5 - 6 | 620 | 62 | |
GenBank mRNA | AB034912.1 | - | 5 - 6 | 576 | 62 |
AK000437.1 | - | 5 - 6 | 591 | 62 | |
AK225525.1 | - | 5 - 6 | 591 | 62 | |
BC086311.1 | - | 5 - 6 | 598 | 62 | |
EF494669.1 | - | 5 - 6 | 497 | 62 |