The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000083.2 | NP_000074.2 | 10 - 11 | 1255 | 75 |
NR_046453.1 | 10 - 11 | 1258 | 75 | ||
XM_017011739.1 | XP_016867228.1 | 7 - 8 | 956 | 75 | |
XM_017011740.1 | XP_016867229.1 | 7 - 8 | 956 | 75 | |
GenBank mRNA | BC112156.1 | - | 10 - 11 | 1220 | 75 |
BC113495.1 | - | 10 - 11 | 1220 | 75 | |
BC143796.1 | - | 10 - 11 | 1223 | 75 | |
M97820.1 | - | 7 - 8 | 659 | 75 | |
Z25884.1 | - | 10 - 11 | 1255 | 75 |