Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRC4
Entrez Gene ID
58484
Gene Name
NLR family CARD domain containing 4
Gene Aliases
AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF
Chromosome Location
Chr.2: 32224449 - 32265743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302504.1 NP_001289433.1 1 - 2 154 92
NM_021209.4 NP_067032.3 1 - 2 154 92
GenBank mRNA AY027787.1 - 1 - 2 155 92
AY027788.1 - 1 - 2 155 92
AY027789.1 - 1 - 2 155 92
AY027790.1 - 1 - 2 155 92
DN998613.1 - 1 - 2 160 92

View Full Product Details