The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199138.1 | NP_001186067.1 | 3 - 4 | 519 | 147 |
NM_001199139.1 | NP_001186068.1 | 3 - 4 | 402 | 147 | |
NM_021209.4 | NP_067032.3 | 3 - 4 | 533 | 147 | |
XM_017004619.1 | XP_016860108.1 | 3 - 4 | 519 | 147 | |
GenBank mRNA | AF376061.1 | - | 4 - 5 | 747 | 147 |
AK095467.1 | - | 3 - 4 | 519 | 147 | |
AK292673.1 | - | 3 - 4 | 504 | 147 | |
AK314762.1 | - | 3 - 4 | 504 | 147 | |
AY027787.1 | - | 3 - 4 | 534 | 147 | |
AY027789.1 | - | 3 - 4 | 534 | 147 | |
AY032589.1 | - | 2 - 3 | 293 | 147 | |
AY035391.1 | - | 3 - 4 | 402 | 147 | |
AY358152.1 | - | 3 - 4 | 481 | 147 | |
BC031555.1 | - | 3 - 4 | 489 | 147 |