The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002928.3 | NP_002919.3 | 4 - 5 | 543 | 91 |
GenBank mRNA | AF493937.1 | - | 4 - 5 | 389 | 91 |
AK311880.1 | - | 4 - 5 | 519 | 91 | |
AW193671.1 | - | 4 - 5 | 538 | 91 | |
BC006243.2 | - | 4 - 5 | 505 | 91 | |
BT006638.1 | - | 4 - 5 | 389 | 91 | |
DA060276.1 | - | 1 - 2 | 261 | 91 | |
U70426.1 | - | 4 - 5 | 481 | 91 | |
U94829.1 | - | 4 - 5 | 392 | 91 |