Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
Chromosome Location
Chr.1: 56929210 - 56974383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278544.1 NP_001265473.1 3 - 4 569 60
GenBank mRNA AK298425.1 - 3 - 4 486 60

View Full Product Details