The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242864.2 | NP_001229793.1 | 7 - 8 | 1020 | 109 |
NM_001535.4 | NP_001526.2 | 9 - 10 | 1326 | 109 | |
NM_206962.3 | NP_996845.1 | 10 - 11 | 1435 | 109 | |
XM_005261111.3 | XP_005261168.1 | 10 - 11 | 1872 | 109 | |
GenBank mRNA | AK095641.1 | - | 2 - 3 | 1629 | 109 |
AK123352.1 | - | 9 - 10 | 2055 | 109 | |
AK123650.1 | - | 10 - 11 | 1863 | 109 | |
AK223157.1 | - | 9 - 10 | 1247 | 109 | |
AK312573.1 | - | 9 - 10 | 1263 | 109 | |
AL833586.1 | - | 9 - 10 | 1253 | 109 | |
BC000727.2 | - | 10 - 11 | 1355 | 109 | |
BC100026.1 | - | 7 - 8 | 934 | 109 | |
CR541804.1 | - | 8 - 9 | 1101 | 109 | |
KU178127.1 | - | 8 - 9 | 1101 | 109 | |
KU178129.1 | - | 5 - 6 | 581 | 109 | |
U80213.1 | - | 9 - 10 | 1266 | 109 | |
X99209.1 | - | 9 - 10 | 1277 | 109 |