The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001535.4 | NP_001526.2 | 7 - 8 | 1051 | 112 |
NM_206962.3 | NP_996845.1 | 8 - 9 | 1160 | 112 | |
XM_005261111.3 | XP_005261168.1 | 8 - 9 | 1597 | 112 | |
GenBank mRNA | AK095641.1 | - | 1 - 2 | 516 | 112 |
AK123352.1 | - | 7 - 8 | 1780 | 112 | |
AK123650.1 | - | 8 - 9 | 1588 | 112 | |
AK124494.1 | - | 1 - 2 | 2090 | 112 | |
AK223157.1 | - | 7 - 8 | 972 | 112 | |
AK312573.1 | - | 7 - 8 | 988 | 112 | |
AL833586.1 | - | 7 - 8 | 978 | 112 | |
BC000727.2 | - | 8 - 9 | 1080 | 112 | |
BC038765.1 | - | 5 - 6 | 1039 | 112 | |
CR541804.1 | - | 6 - 7 | 826 | 112 | |
KU178127.1 | - | 6 - 7 | 826 | 112 | |
KU178129.1 | - | 3 - 4 | 306 | 112 | |
U80213.1 | - | 7 - 8 | 991 | 112 | |
X99209.1 | - | 7 - 8 | 1002 | 112 |