Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LANCL2
Entrez Gene ID
55915
Gene Name
LanC like 2
Gene Aliases
GPR69B, TASP
Chromosome Location
Chr.7: 55365448 - 55433742 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018697.3 NP_061167.1 8 - 9 1840 112
GenBank mRNA AB035966.1 - 8 - 9 1317 112
AF353942.1 - 8 - 9 1317 112
AJ278245.1 - 8 - 9 1448 112
AK095857.1 - 8 - 9 1840 112
AK225958.1 - 8 - 9 1293 112
AK301747.1 - 8 - 9 1246 112
AK313326.1 - 8 - 9 1585 112
AL713665.2 - 8 - 9 1232 112
BC004887.2 - 5 - 6 668 112
BC070049.1 - 8 - 9 1536 112

View Full Product Details