Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CABIN1
Entrez Gene ID
23523
Gene Name
calcineurin binding protein 1
Gene Aliases
CAIN, KB-318B8.7, PPP3IN
Chromosome Location
Chr.22: 24011192 - 24178628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199281.1 NP_001186210.1 24 - 25 4221 67
NM_001201429.1 NP_001188358.1 23 - 24 3765 67
NM_012295.3 NP_036427.1 24 - 25 3915 67
XM_005261415.2 XP_005261472.1 23 - 24 3862 67
XM_005261416.2 XP_005261473.1 23 - 24 3862 67
XM_005261417.2 XP_005261474.1 23 - 24 3862 67
XM_005261419.2 XP_005261476.1 23 - 24 3903 67
XM_006724179.1 XP_006724242.1 23 - 24 3862 67
XM_006724180.1 XP_006724243.1 23 - 24 3862 67
XM_006724181.1 XP_006724244.1 23 - 24 3862 67
XM_011530026.1 XP_011528328.1 22 - 23 3667 67
XM_011530029.2 XP_011528331.1 23 - 24 3862 67
XM_011530030.2 XP_011528332.1 23 - 24 3862 67
XM_017028680.1 XP_016884169.1 23 - 24 3862 67
XM_017028681.1 XP_016884170.1 22 - 23 3712 67
XM_017028682.1 XP_016884171.1 23 - 24 3783 67
XM_017028683.1 XP_016884172.1 22 - 23 3712 67
XM_017028684.1 XP_016884173.1 8 - 9 1400 67
GenBank mRNA AB002328.2 - 24 - 25 4138 67
AF072441.1 - 24 - 25 3916 67
BC054497.1 - 23 - 24 3732 67

View Full Product Details