Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC17A8
Entrez Gene ID
246213
Gene Name
solute carrier family 17 member 8
Gene Aliases
DFNA25, VGLUT3
Chromosome Location
Chr.12: 100357079 - 100422059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145288.1 NP_001138760.1 4 - 5 898 59
NM_139319.2 NP_647480.1 4 - 5 898 59
GenBank mRNA AJ459241.1 - 4 - 5 761 59
AK128319.1 - 4 - 5 898 59
BC117229.1 - 4 - 5 609 59
BC143396.1 - 4 - 5 609 59

View Full Product Details