The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022351.4 | NP_071746.1 | 8 - 9 | 893 | 135 |
XM_011517213.2 | XP_011515515.1 | 8 - 9 | 818 | 135 | |
GenBank mRNA | AF193756.1 | - | 4 - 5 | 411 | 135 |
AF414126.2 | - | 9 - 10 | 982 | 135 | |
AK095533.1 | - | 8 - 9 | 806 | 135 | |
AY224210.1 | - | 8 - 9 | 699 | 135 | |
BC068448.1 | - | 8 - 9 | 806 | 135 |