The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000562.2 | NP_000553.1 | 10 - 11 | 1741 | 69 |
GenBank mRNA | BC132911.1 | - | 10 - 11 | 1633 | 69 |
BC132913.1 | - | 10 - 11 | 1633 | 69 | |
M16974.1 | - | 10 - 11 | 1746 | 69 |