The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135653.1 | NP_001129125.1 | 10 - 11 | 2350 | 73 |
GenBank mRNA | AB208962.1 | - | 4 - 5 | 546 | 73 |
AK222917.1 | - | 4 - 5 | 587 | 73 | |
BC029969.1 | - | 10 - 11 | 1902 | 73 | |
BC071591.1 | - | 10 - 11 | 2288 | 73 | |
BC094755.1 | - | 10 - 11 | 2291 | 73 | |
BC106908.1 | - | 5 - 6 | 730 | 73 |