The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135653.1 | NP_001129125.1 | 3 - 4 | 1408 | 63 |
NM_001135654.1 | NP_001129126.1 | 3 - 4 | 1408 | 63 | |
NM_003819.3 | NP_003810.1 | 3 - 4 | 1408 | 63 | |
GenBank mRNA | BC029969.1 | - | 3 - 4 | 960 | 63 |
BC065540.1 | - | 3 - 4 | 1311 | 63 | |
BC071591.1 | - | 3 - 4 | 1346 | 63 | |
BC094755.1 | - | 3 - 4 | 1349 | 63 | |
U33818.1 | - | 3 - 4 | 663 | 63 | |
U75686.1 | - | 3 - 4 | 1304 | 63 |