The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135653.1 | NP_001129125.1 | 4 - 5 | 1535 | 75 |
NM_001135654.1 | NP_001129126.1 | 4 - 5 | 1535 | 75 | |
NM_003819.3 | NP_003810.1 | 4 - 5 | 1535 | 75 | |
GenBank mRNA | BC029969.1 | - | 4 - 5 | 1087 | 75 |
BC065540.1 | - | 4 - 5 | 1438 | 75 | |
BC071591.1 | - | 4 - 5 | 1473 | 75 | |
BC094755.1 | - | 4 - 5 | 1476 | 75 | |
D12799.1 | - | 1 - 2 | 70 | 75 | |
U33818.1 | - | 4 - 5 | 790 | 75 | |
U75686.1 | - | 4 - 5 | 1431 | 75 |