The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001621.4 | NP_001612.1 | 1 - 2 | 675 | 84 |
GenBank mRNA | AK307495.1 | - | 1 - 2 | 675 | 84 |
BC069390.1 | - | 1 - 2 | 132 | 84 | |
BC070080.1 | - | 1 - 2 | 593 | 84 | |
D16354.1 | - | 1 - 2 | 644 | 84 | |
L19872.1 | - | 1 - 2 | 437 | 84 |