The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000417.2 | NP_000408.1 | 5 - 6 | 872 | 71 |
NM_001308242.1 | NP_001295171.1 | 4 - 5 | 656 | 71 | |
GenBank mRNA | AK223313.1 | - | 5 - 6 | 794 | 71 |
AK313847.1 | - | 5 - 6 | 869 | 71 | |
K03122.1 | - | 4 - 5 | 596 | 71 | |
M14098.1 | - | 4 - 5 | 590 | 71 | |
X01057.1 | - | 5 - 6 | 833 | 71 |