Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF12
Entrez Gene ID
113220
Gene Name
kinesin family member 12
Gene Aliases
-
Chromosome Location
Chr.9: 114091623 - 114100099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005251683.4 XP_005251740.1 3 - 4 981 129

View Full Product Details