Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
Chromosome Location
Chr.10: 6010694 - 6062370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308242.1 NP_001295171.1 3 - 4 586 56
GenBank mRNA K03122.1 - 3 - 4 526 56

View Full Product Details