The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170414.2 | NP_001163885.1 | 14 - 15 | 2157 | 76 |
NM_001170415.1 | NP_001163886.1 | 13 - 14 | 2157 | 76 | |
NM_001170416.1 | NP_001163887.1 | 13 - 14 | 2087 | 76 | |
NM_004817.3 | NP_004808.2 | 13 - 14 | 2312 | 76 | |
NM_201629.3 | NP_963923.1 | 13 - 14 | 2312 | 76 | |
XM_005252314.2 | XP_005252371.1 | 13 - 14 | 2365 | 76 | |
XM_011519204.1 | XP_011517506.1 | 13 - 14 | 2055 | 76 | |
XM_011519206.2 | XP_011517508.1 | 15 - 16 | 2657 | 76 | |
XM_011519207.2 | XP_011517509.1 | 14 - 15 | 2153 | 76 | |
XM_011519208.2 | XP_011517510.1 | 14 - 15 | 2113 | 76 | |
XM_011519209.2 | XP_011517511.1 | 12 - 13 | 1942 | 76 | |
XM_017015326.1 | XP_016870815.1 | 14 - 15 | 2210 | 76 | |
XM_017015327.1 | XP_016870816.1 | 13 - 14 | 2312 | 76 | |
XM_017015328.1 | XP_016870817.1 | 13 - 14 | 2365 | 76 | |
GenBank mRNA | AF489824.1 | - | 13 - 14 | 2073 | 76 |
AK295016.1 | - | 14 - 15 | 2113 | 76 | |
AK295034.1 | - | 13 - 14 | 2087 | 76 | |
AK302483.1 | - | 13 - 14 | 76 | ||
AK304447.1 | - | 15 - 16 | 2239 | 76 | |
BC027592.1 | - | 13 - 14 | 2203 | 76 | |
L27476.1 | - | 13 - 14 | 2073 | 76 |