Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDR
Entrez Gene ID
3791
Gene Name
kinase insert domain receptor
Gene Aliases
CD309, FLK1, VEGFR, VEGFR2
Chromosome Location
Chr.4: 55078259 - 55125595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002253.2 NP_002244.1 12 - 13 1950 75
GenBank mRNA AB209901.1 - 12 - 13 1935 75
AF035121.1 - 12 - 13 1951 75
AF063658.1 - 12 - 13 1648 75
BC131822.1 - 12 - 13 1710 75
EU826563.1 - 12 - 13 1648 75
FJ899739.1 - 12 - 13 1648 75
L04947.1 - 12 - 13 1642 75
X61656.1 - 12 - 13 1642 75

View Full Product Details