The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000917.3 | NP_000908.2 | 11 - 12 | 1541 | 67 |
NM_001017962.2 | NP_001017962.1 | 11 - 12 | 1541 | 67 | |
NM_001142595.1 | NP_001136067.1 | 12 - 13 | 1634 | 67 | |
GenBank mRNA | AK222960.1 | - | 11 - 12 | 1429 | 67 |
AK312593.1 | - | 11 - 12 | 1427 | 67 | |
BC034998.1 | - | 11 - 12 | 1434 | 67 | |
CD013930.1 | - | 12 - 13 | 1513 | 67 | |
M24486.1 | - | 11 - 12 | 1418 | 67 | |
M24487.1 | - | 11 - 12 | 1418 | 67 |