The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022366.2 | NP_071761.1 | 2 - 3 | 524 | 151 |
GenBank mRNA | AF529366.1 | - | 2 - 3 | 399 | 151 |
AK026835.1 | - | 2 - 3 | 523 | 151 | |
BC003383.1 | - | 2 - 3 | 510 | 151 |