Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
Chromosome Location
Chr.2: 237324012 - 237434822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004369.3 NP_004360.2 13 - 14 6203 72
NM_057166.4 NP_476507.3 10 - 11 4382 72
NM_057167.3 NP_476508.2 12 - 13 5585 72
XM_005246065.1 XP_005246122.1 12 - 13 5560 72
XM_005246066.1 XP_005246123.1 10 - 11 4706 72
XM_006712253.1 XP_006712316.1 13 - 14 5659 72
XM_011510574.1 XP_011508876.1 13 - 14 6157 72
XM_017003303.1 XP_016858792.1 18 - 19 6210 72
XM_017003304.1 XP_016858793.1 15 - 16 4422 72
GenBank mRNA AB209636.1 - 4 - 5 1494 72
BC144595.1 - 10 - 11 4323 72
BC150625.1 - 10 - 11 4323 72
BC171790.1 - 10 - 11 4323 72
X52022.1 - 13 - 14 6173 72

View Full Product Details