Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
Chromosome Location
Chr.2: 237324012 - 237434822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004369.3 NP_004360.2 16 - 17 6495 81
NM_057166.4 NP_476507.3 13 - 14 4674 81
NM_057167.3 NP_476508.2 15 - 16 5877 81
XM_005246065.1 XP_005246122.1 15 - 16 5852 81
XM_005246066.1 XP_005246123.1 13 - 14 4998 81
XM_006712253.1 XP_006712316.1 16 - 17 5951 81
XM_011510574.1 XP_011508876.1 16 - 17 6449 81
XM_017003303.1 XP_016858792.1 21 - 22 6502 81
XM_017003304.1 XP_016858793.1 18 - 19 4714 81
GenBank mRNA AB209636.1 - 7 - 8 1786 81
BC144595.1 - 13 - 14 4615 81
BC150625.1 - 13 - 14 4615 81
BC171790.1 - 13 - 14 4615 81
M20778.1 - 2 - 3 99 81
X52022.1 - 16 - 17 6465 81

View Full Product Details