Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
Chromosome Location
Chr.2: 237324012 - 237434822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004369.3 NP_004360.2 27 - 28 7107 123
NM_057166.4 NP_476507.3 24 - 25 5286 123
NM_057167.3 NP_476508.2 26 - 27 6489 123
XM_005246065.1 XP_005246122.1 26 - 27 6464 123
XM_005246066.1 XP_005246123.1 24 - 25 5610 123
XM_006712253.1 XP_006712316.1 27 - 28 6563 123
XM_011510574.1 XP_011508876.1 27 - 28 7061 123
XM_017003303.1 XP_016858792.1 32 - 33 7114 123
XM_017003304.1 XP_016858793.1 29 - 30 5326 123
GenBank mRNA AB209636.1 - 18 - 19 2398 123
BC033174.1 - 4 - 5 230 123
BC144595.1 - 24 - 25 5227 123
BC150625.1 - 24 - 25 5227 123
BC171790.1 - 24 - 25 5227 123
BX641155.1 - 4 - 5 1076 123
M20778.1 - 13 - 14 711 123
X52022.1 - 27 - 28 7077 123

View Full Product Details