Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
Chromosome Location
Chr.2: 237324012 - 237434822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004369.3 NP_004360.2 39 - 40 8857 87
NM_057166.4 NP_476507.3 36 - 37 7036 87
NM_057167.3 NP_476508.2 38 - 39 8239 87
XM_005246065.1 XP_005246122.1 38 - 39 8214 87
XM_005246066.1 XP_005246123.1 36 - 37 7360 87
XM_006712253.1 XP_006712316.1 39 - 40 8313 87
XM_011510574.1 XP_011508876.1 39 - 40 8811 87
XM_017003303.1 XP_016858792.1 44 - 45 8864 87
XM_017003304.1 XP_016858793.1 41 - 42 7076 87
GenBank mRNA AB209636.1 - 30 - 31 4148 87
BC033174.1 - 16 - 17 1980 87
BC144595.1 - 36 - 37 6977 87
BC150625.1 - 36 - 37 6977 87
BC171790.1 - 36 - 37 6977 87
BX641155.1 - 16 - 17 4974 87
X52022.1 - 39 - 40 8827 87

View Full Product Details