The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016037.3 | NP_057121.2 | 4 - 5 | 432 | 107 |
XM_011541555.2 | XP_011539857.1 | 3 - 4 | 366 | 107 | |
GenBank mRNA | AF067802.1 | - | 4 - 5 | 395 | 107 |
AF151852.1 | - | 4 - 5 | 410 | 107 | |
AK023587.1 | - | 2 - 3 | 1301 | 107 | |
AK291900.1 | - | 4 - 5 | 362 | 107 | |
AK296018.1 | - | 4 - 5 | 365 | 107 | |
AK308377.1 | - | 3 - 4 | 367 | 107 | |
BC005182.1 | - | 4 - 5 | 377 | 107 | |
BU940926.1 | - | 4 - 5 | 435 | 107 |