Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFB2M
Entrez Gene ID
64216
Gene Name
transcription factor B2, mitochondrial
Gene Aliases
Hkp1, mtTFB2
Chromosome Location
Chr.1: 246540561 - 246566263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017002055.1 XP_016857544.1 2 - 3 468 87
GenBank mRNA AK300238.1 - 2 - 3 526 87

View Full Product Details