The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007277.4 | NP_009208.2 | 11 - 12 | 2063 | 63 |
XM_017009003.1 | XP_016864492.1 | 11 - 12 | 2098 | 63 | |
XM_017009004.1 | XP_016864493.1 | 12 - 13 | 2176 | 63 | |
XM_017009005.1 | XP_016864494.1 | 8 - 9 | 1747 | 63 | |
XM_017009006.1 | XP_016864495.1 | 10 - 11 | 1991 | 63 | |
GenBank mRNA | AF055006.1 | - | 8 - 9 | 1115 | 63 |
AK074086.1 | - | 6 - 7 | 2629 | 63 | |
AK131103.1 | - | 7 - 8 | 2242 | 63 | |
AK315749.1 | - | 8 - 9 | 1571 | 63 | |
AL832434.1 | - | 9 - 10 | 1719 | 63 | |
BC001511.2 | - | 8 - 9 | 1475 | 63 | |
BC019304.1 | - | 9 - 10 | 1645 | 63 | |
BC064569.1 | - | 10 - 11 | 2005 | 63 | |
GU727644.1 | - | 11 - 12 | 2063 | 63 |