The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011885.1 | NP_001011885.1 | 4 - 5 | 1065 | 75 |
NM_025238.3 | NP_079514.1 | 4 - 5 | 1065 | 75 | |
GenBank mRNA | AF257241.1 | - | 4 - 5 | 944 | 75 |
AF355402.1 | - | 4 - 5 | 945 | 75 | |
AF529369.1 | - | 4 - 5 | 862 | 75 | |
AK000731.1 | - | 4 - 5 | 929 | 75 | |
AK225355.1 | - | 4 - 5 | 929 | 75 | |
AK225362.1 | - | 4 - 5 | 929 | 75 | |
AK294410.1 | - | 2 - 3 | 317 | 75 | |
AK298534.1 | - | 1 - 2 | 212 | 75 | |
AL136853.1 | - | 4 - 5 | 945 | 75 | |
AY364253.1 | - | 4 - 5 | 862 | 75 | |
BC017464.1 | - | 4 - 5 | 892 | 75 | |
BC028097.1 | - | 4 - 5 | 965 | 75 |