The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017962.2 | NP_001017962.1 | 9 - 10 | 1390 | 68 |
NM_001142595.1 | NP_001136067.1 | 10 - 11 | 1483 | 68 | |
GenBank mRNA | BC034998.1 | - | 9 - 10 | 1283 | 68 |
CD013930.1 | - | 10 - 11 | 1362 | 68 |