Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6K3
Entrez Gene ID
391114
Gene Name
olfactory receptor family 6 subfamily K member 3
Gene Aliases
OR1-18
Chromosome Location
Chr.1: 158717168 - 158718115 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005327.2 NP_001005327.2 1 - 1 887 104

View Full Product Details