Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- IMP3
- Entrez Gene ID
-
55272
- Gene Name
- IMP3, U3 small nucleolar ribonucleoprotein
- Gene Aliases
-
BRMS2, C15orf12, MRPS4
- Chromosome Location
-
Chr.15: 75639085 - 75640323 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK057531.1 |
- |
1 - 2 |
843 |
91 |
View Full Product Details