Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLD2
Entrez Gene ID
5425
Gene Name
DAN polymerase delta 2, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.7: 44114680 - 44123668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127218.2 NP_001120690.1 7 - 8 1148 77
NM_001256879.1 NP_001243808.1 8 - 9 1509 77
NM_006230.3 NP_006221.2 7 - 8 975 77
XM_011515431.1 XP_011513733.1 7 - 8 971 77
GenBank mRNA AK292347.1 - 8 - 9 1509 77
AK298168.1 - 7 - 8 839 77
AK312294.1 - 7 - 8 1108 77
BC000459.2 - 7 - 8 938 77
BC030001.2 - 6 - 7 915 77
U21090.1 - 7 - 8 937 77

View Full Product Details