Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLD2
Entrez Gene ID
5425
Gene Name
DAN polymerase delta 2, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.7: 44114680 - 44123668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127218.2 NP_001120690.1 9 - 10 1432 79
NM_001256879.1 NP_001243808.1 10 - 11 1793 79
NM_006230.3 NP_006221.2 9 - 10 1259 79
XM_011515431.1 XP_011513733.1 9 - 10 1255 79
GenBank mRNA AK292347.1 - 10 - 11 1793 79
AK298168.1 - 9 - 10 1123 79
AK312294.1 - 9 - 10 1392 79
BC000459.2 - 9 - 10 1222 79
BC030001.2 - 8 - 9 1199 79

View Full Product Details