The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000670.4 | NP_000661.2 | 1 - 2 | 91 | 71 |
XM_017007715.1 | XP_016863204.1 | 1 - 2 | 123 | 71 | |
GenBank mRNA | AK290835.1 | - | 1 - 2 | 90 | 71 |
AK313649.1 | - | 1 - 2 | 90 | 71 | |
AV659353.1 | - | 1 - 2 | 91 | 71 | |
AV659364.1 | - | 1 - 2 | 91 | 71 | |
BC022319.1 | - | 1 - 2 | 90 | 71 | |
EU668317.1 | - | 1 - 2 | 76 | 71 | |
M15943.1 | - | 1 - 2 | 76 | 71 |