The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000670.4 | NP_000661.2 | 5 - 6 | 657 | 73 |
NM_001306171.1 | NP_001293100.1 | 6 - 7 | 805 | 73 | |
NM_001306172.1 | NP_001293101.1 | 6 - 7 | 797 | 73 | |
XM_017007713.1 | XP_016863202.1 | 6 - 7 | 828 | 73 | |
XM_017007714.1 | XP_016863203.1 | 6 - 7 | 836 | 73 | |
XM_017007715.1 | XP_016863204.1 | 5 - 6 | 689 | 73 | |
GenBank mRNA | AK290835.1 | - | 5 - 6 | 656 | 73 |
AK295556.1 | - | 6 - 7 | 804 | 73 | |
AK313649.1 | - | 5 - 6 | 656 | 73 | |
BC022319.1 | - | 5 - 6 | 656 | 73 | |
CD014080.1 | - | 6 - 7 | 775 | 73 | |
EU668317.1 | - | 5 - 6 | 642 | 73 | |
M15943.1 | - | 5 - 6 | 642 | 73 |