The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000218.2 | NP_000209.2 | 10 - 11 | 1506 | 126 |
NM_181798.1 | NP_861463.1 | 10 - 11 | 1273 | 126 | |
GenBank mRNA | AF000571.1 | - | 10 - 11 | 1508 | 126 |
AF051426.1 | - | 10 - 11 | 1270 | 126 | |
AK129678.1 | - | 8 - 9 | 1065 | 126 | |
AK290618.1 | - | 10 - 11 | 1264 | 126 | |
AY114213.1 | - | 10 - 11 | 1398 | 126 | |
BC017074.1 | - | 5 - 6 | 547 | 126 | |
BC113545.1 | - | 10 - 11 | 1073 | 126 | |
U89364.1 | - | 10 - 11 | 1200 | 126 |