The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144888.1 | NP_001138360.1 | 10 - 11 | 1379 | 71 |
NM_006340.2 | NP_006331.1 | 10 - 11 | 1379 | 71 | |
NM_017450.2 | NP_059344.1 | 10 - 11 | 1379 | 71 | |
NM_017451.2 | NP_059345.1 | 10 - 11 | 1379 | 71 | |
XM_005256943.1 | XP_005257000.1 | 11 - 12 | 1478 | 71 | |
XM_005256944.1 | XP_005257001.1 | 11 - 12 | 1478 | 71 | |
XM_005256945.1 | XP_005257002.1 | 11 - 12 | 1478 | 71 | |
XM_005256948.3 | XP_005257005.1 | 10 - 11 | 1382 | 71 | |
XM_006721635.1 | XP_006721698.1 | 11 - 12 | 1481 | 71 | |
XM_006721636.1 | XP_006721699.1 | 11 - 12 | 1481 | 71 | |
XM_006721637.2 | XP_006721700.1 | 10 - 11 | 1382 | 71 | |
XM_011524193.1 | XP_011522495.1 | 11 - 12 | 1481 | 71 | |
XM_011524194.1 | XP_011522496.1 | 11 - 12 | 1481 | 71 | |
XM_011524195.1 | XP_011522497.1 | 5 - 6 | 1141 | 71 | |
XM_011524196.1 | XP_011522498.1 | 5 - 6 | 903 | 71 | |
XM_011524197.1 | XP_011522499.1 | 5 - 6 | 900 | 71 | |
XM_017024017.1 | XP_016879506.1 | 9 - 10 | 2139 | 71 | |
XM_017024018.1 | XP_016879507.1 | 8 - 9 | 1601 | 71 | |
XM_017024019.1 | XP_016879508.1 | 5 - 6 | 1395 | 71 | |
GenBank mRNA | AB015019.1 | - | 10 - 11 | 1364 | 71 |
AB015020.1 | - | 10 - 11 | 1364 | 71 | |
AB017119.1 | - | 10 - 11 | 1364 | 71 | |
AB017120.1 | - | 10 - 11 | 1364 | 71 | |
AK023688.1 | - | 11 - 12 | 1464 | 71 | |
AK056888.1 | - | 6 - 7 | 1551 | 71 | |
AK225612.1 | - | 10 - 11 | 1376 | 71 | |
AK297644.1 | - | 10 - 11 | 1308 | 71 | |
AK298629.1 | - | 8 - 9 | 1214 | 71 | |
AK301436.1 | - | 5 - 6 | 860 | 71 | |
BC002495.2 | - | 10 - 11 | 1354 | 71 | |
BC014020.2 | - | 10 - 11 | 1369 | 71 | |
BC032559.1 | - | 10 - 11 | 1377 | 71 |