The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025108.2 | NP_079384.2 | 8 - 9 | 1001 | 73 |
XM_011522667.1 | XP_011520969.1 | 7 - 8 | 905 | 73 | |
GenBank mRNA | AK023971.1 | - | 7 - 8 | 1082 | 73 |
AK093788.1 | - | 7 - 8 | 1239 | 73 | |
AK128408.1 | - | 6 - 7 | 1831 | 73 | |
AK301929.1 | - | 8 - 9 | 997 | 73 | |
BC018719.1 | - | 8 - 9 | 989 | 73 | |
CR457319.1 | - | 5 - 6 | 469 | 73 |