Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAD1
Entrez Gene ID
132612
Gene Name
adenosine deaminase domain containing 1
Gene Aliases
Tenr
Chromosome Location
Chr.4: 122378966 - 122429815 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159285.1 NP_001152757.1 5 - 6 752 165
NM_001159295.1 NP_001152767.1 5 - 6 816 165
NM_139243.3 NP_640336.1 6 - 7 830 165
XM_005262741.2 XP_005262798.1 5 - 6 851 165
XM_005262743.2 XP_005262800.1 5 - 6 751 165
XM_005262744.2 XP_005262801.1 5 - 6 851 165
XM_011531603.1 XP_011529905.1 5 - 6 749 165
GenBank mRNA AK057303.1 - 6 - 7 784 165
BC040229.1 - 5 - 6 698 165

View Full Product Details