The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145721.1 | NP_001139193.1 | 7 - 8 | 793 | 103 |
NM_001145722.1 | NP_001139194.1 | 7 - 8 | 1215 | 103 | |
NM_001145724.1 | NP_001139196.1 | 5 - 6 | 653 | 103 | |
NM_004838.3 | NP_004829.3 | 7 - 8 | 943 | 103 | |
NR_027297.1 | 6 - 7 | 1347 | 103 | ||
XM_006722943.1 | XP_006723006.1 | 7 - 8 | 827 | 103 | |
XM_006722944.1 | XP_006723007.1 | 7 - 8 | 845 | 103 | |
GenBank mRNA | AF093265.1 | - | 7 - 8 | 784 | 103 |
AK124450.1 | - | 6 - 7 | 1347 | 103 | |
AK313995.1 | - | 7 - 8 | 1216 | 103 | |
BC012113.1 | - | 7 - 8 | 885 | 103 | |
BM915739.1 | - | - | 300 | 103 | |
BT019464.1 | - | 6 - 7 | 694 | 103 | |
Y17573.1 | - | 6 - 7 | 694 | 103 | |
Y18894.1 | - | 5 - 6 | 586 | 103 |